Learn More
Electron Microscopy Sciences Tissue Matrice Spinal Cord Coronal, 0.5 mm Rat, 4 mm W x 96 mm L Cavity, 5 mm Depth
SDP
Catalog No. NC1625606
Encompass_Preferred
These spinal cord matrices are precisely machined to ensure reproducible 0.5mm or 1mm sections of mouse or rat spinal cords.
Type: 0.5 mm Rat
Cavity: 4 mm W x 96 mm L
Depth: 5 mm
Catalog No. NC1625606 Supplier Electron Microscopy Sciences Supplier No. 69084C
These spinal cord matrices are precisely machined to ensure reproducible 0.5mm or 1mm sections of mouse or rat spinal cords.
Type: 0.5 mm Rat
Cavity: 4 mm W x 96 mm L
Depth: 5 mm