Learn More
Biomatik Corporation Rat Spleen Focus Forming Virus Proviral Integration Oncogene (SPI1) ELISA Kit
Supplier:  Biomatik Corporation EKU07441
Encompass_Preferred
Rat Spleen Focus Forming Virus Proviral Integration Oncogene (SPI1) ELISA Kit for detecting SPI1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6BDS1. Gene ID: 366126. Target Synonyms: OF; PU.1; SFPI1; SPI-1; SPI-A; SPI-1 Proto-Oncogene; Hematopoietic Transcription Factor PU.1; 31 kDa Transforming Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6811