Learn More
Innovative Research Inc Rat Spleen Tss Lysate 100ug
Catalog No. 502743995
Encompass_Preferred
Rat Spleen Tissue Lysate 100ug - Rat Spleen Tissue Lysate from Innovative Research is provided as a Liquid buffered in SDS sample buffer containing 5% beta-mercaptoethanol. This product is especially useful for applications like Western Blot, among others. For continuous use, store at 2-8C. If not using the entire volume immediately, store single-use aliquots at -70C. Repeated freeze/thaw cycles are not recommended. Under proper storage conditions, the shelf life is six months from the date of receipt. Additional Details Species Rat Source Spleen tissue lysate Format Liquid Storage Conditions -70C
Catalog No. 50-274-3995 Supplier Innovative Research Inc Supplier No. IRTSPTL100UG
Rat Spleen Tissue Lysate 100ug - Rat Spleen Tissue Lysate from Innovative Research is provided as a Liquid buffered in SDS sample buffer containing 5% beta-mercaptoethanol. This product is especially useful for applications like Western Blot, among others. For continuous use, store at 2-8C. If not using the entire volume immediately, store single-use aliquots at -70C. Repeated freeze/thaw cycles are not recommended. Under proper storage conditions, the shelf life is six months from the date of receipt. Additional Details Species Rat Source Spleen tissue lysate Format Liquid Storage Conditions -70C