Learn More
Lonza Walkersville Rat Sprague Dawley Hepatocytes, Cryopreserved, Plateable
Catalog No. NC1885857
Encompass
Cryopreserved Sprague Dawley Rat Hepatocytes, plateable qualifiedare specifically pre-tested for metabolic activity of general Phase I and Phase II metabolism in suspension. They are tested for healthy morphology lasting 72 hours in monolayer sandwich culture.Many batches are talso ested for spheroid formation capacity.Look for the Verified for Spheroidscharacteristic on each CofA.Benefits:Plateable>5Million cells/vialCharacterized for general drug metabolismApplications:Drug metabolism in plated monolayer formatspecies-species comparisonsgenomicsContent:Cryo vial, > 5 million viable cells
Catalog No. NC1885857 Supplier Lonza Walkersville Supplier No. RSCP01
Cryopreserved Sprague Dawley Rat Hepatocytes, plateable qualifiedare specifically pre-tested for metabolic activity of general Phase I and Phase II metabolism in suspension. They are tested for healthy morphology lasting 72 hours in monolayer sandwich culture.Many batches are talso ested for spheroid formation capacity.Look for the Verified for Spheroidscharacteristic on each CofA.Benefits:Plateable>5Million cells/vialCharacterized for general drug metabolismApplications:Drug metabolism in plated monolayer formatspecies-species comparisonsgenomicsContent:Cryo vial, > 5 million viable cells