Learn More
Lonza Walkersville Rat Sprague Dawley Heps Susp
Catalog No. 5000496243
Encompass
Cryopreserved Sprague Dawley rat Hepatocytes suspension are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension using ECOD outputs 7EC formation and formation of 7EC SULT and UGT products This product is not suitable for applications requiring adherant cells
Catalog No. 50-004-96243 Supplier Lonza Walkersville Supplier No. RSCS01
Cryopreserved Sprague Dawley rat Hepatocytes suspension are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension using ECOD outputs 7EC formation and formation of 7EC SULT and UGT products This product is not suitable for applications requiring adherant cells