Learn More
Gibco™ Rat (Sprague Dawley) Microsomes
Description
Liver microsomes prepared from a pool of male Sprague Dawley rats.
Total protein concentration: 20 mg/ml
We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.
Related Links:
Specifications
Specifications
| Concentration | 20 mg/mL |
| Product Type | Subcellular Fractions |
| Content And Storage | Store between -60°C and -85°C |
| Donor Source | Pooled |
| Gender | Male |
| Shipping Condition | Dry Ice |
| Sub Cellular Localization | Microsomes |
| Target Species | Rat |
| Tissue | Liver |
| Form | Frozen |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.