Learn More
Innovative Research Inc Rat Sprg Dwly Albumin 200mg
Catalog No. 502743997
Encompass_Preferred
Rat Sprague Dawley Albumin 200mg - Rat Sprague Dawley Albumin from Innovative Research has been purified from US origin Rat Plasma using proprietary chromatographic and cold ethanol fractionation methods. This is a salt free lyophilized powder with a purity of >95%. This product is useful as a positive control for ELISA and western blot and for a variety of in vitro and in vivo applications including blocking, cell signaling, drug studies, protein-protein interactions, protein stabilization, receptor binding, and tissue culture. This is an ultra-pure preparation, especially as compared to commercially available heat-shock preparations of Albumin, which are less pure and may contain denatured albumin.
Catalog No. 50-274-3997 Supplier Innovative Research Inc Supplier No. IRTSDALB200MG
Rat Sprague Dawley Albumin 200mg - Rat Sprague Dawley Albumin from Innovative Research has been purified from US origin Rat Plasma using proprietary chromatographic and cold ethanol fractionation methods. This is a salt free lyophilized powder with a purity of >95%. This product is useful as a positive control for ELISA and western blot and for a variety of in vitro and in vivo applications including blocking, cell signaling, drug studies, protein-protein interactions, protein stabilization, receptor binding, and tissue culture. This is an ultra-pure preparation, especially as compared to commercially available heat-shock preparations of Albumin, which are less pure and may contain denatured albumin.