Learn More
Bioworld RAT STEM CELL FACTOR (RECOM
Supplier:  Bioworld 220606751
Encompass_Preferred
RAT STEM CELL FACTOR (RECOMBINANT): Recombinant Rat Stem Cell Factor (SCF) Source: E.Coli Description: Recombinant Rat Stem Cell Factor (SCF) produced in E.Coli is a single, non-glycosylated polypeptide chain containing 165 amino aci...
Catalog No. 50-253-771