Learn More
Biomatik Corporation Rat Stem Cell Factor (SCF) ELISA Kit
Supplier:  Biomatik Corporation EKU07466
Encompass_Preferred
Rat Stem Cell Factor (SCF) ELISA Kit for detecting SCF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21581. Gene ID: 60427. Target Synonyms: KITLG; MGF; SF; KL1; Kitl; FPH2; Kit-Ligand; Steel Factor; Mast Cell Growth Factor; Steel factor; Familial Progressive Hyperpigmentation 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7422