Learn More
Euthanex Rat Stereotaxic Breather
Supplier:  Euthanex EZ107A
Encompass_Preferred
Rat Stereotaxic Breather, specifically designed for use with rats, provides precise stabilization of head position and includes adjustable nose bridge, lower jaw support and ear bar. Slips over toothbar of any rat stereotaxic adapter.
Catalog No. 50-154-0147