Learn More
American Research Products Inc Rat Steroid 5 Alpha Reductase
Catalog No. 5000330225
Encompass
Rat Steroid 5-Alpha-Reductase 1 (SRD5A1) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-30225 Supplier American Research Products Inc Supplier No. CSBE14053R
Rat Steroid 5-Alpha-Reductase 1 (SRD5A1) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich