Learn More
Cayman Chemical Rat Estrogen Receptor Alpha Reporter Assay System, 3x32 Well
SDP
Catalog No. 502913893
Encompass_Preferred
rERα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. R00401-32, R00401]
Catalog No. 50-291-3893 Supplier Cayman Chemical Supplier No. 367093X32 WELL
rERα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. R00401-32, R00401]