Learn More
American Research Products Inc Rat Stromal cell deriVed facto
Catalog No. 5000328661
Encompass
Rat Stromal cell deriVed factor 1a SDF-1a ELISA Kit from CUSABIO Detection Range 0 78 pg/mL-50 pg/mL Sensitivity 0 195 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28661 Supplier American Research Products Inc Supplier No. CSBE08729R
Rat Stromal cell deriVed factor 1a SDF-1a ELISA Kit from CUSABIO Detection Range 0 78 pg/mL-50 pg/mL Sensitivity 0 195 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich