Learn More
American Research Products Inc Rat Sulfotransferase Family 1E
Catalog No. 5000329431
Encompass
Rat Sulfotransferase Family 1E Member 1 (SULT1E1) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29431 Supplier American Research Products Inc Supplier No. CSBE13472R
Rat Sulfotransferase Family 1E Member 1 (SULT1E1) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich