Learn More
American Research Products Inc Rat superoXide dismutase 1 so
Catalog No. 5000327672
Encompass
Rat superoXide dismutase 1 soluble (SOD1) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27672 Supplier American Research Products Inc Supplier No. CSBEL022397RA
Rat superoXide dismutase 1 soluble (SOD1) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich