Learn More
American Research Products Inc Rat SuperoXide dismutase Mn
Catalog No. 5000327538
Encompass
Rat SuperoXide dismutase [Mn] mitochondrial (SOD2) ELISA kit from CUSABIO Detection Range 1 87 pg/mL-120 pg/mL Sensitivity 0 47 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27538 Supplier American Research Products Inc Supplier No. CSBEL022398RA
Rat SuperoXide dismutase [Mn] mitochondrial (SOD2) ELISA kit from CUSABIO Detection Range 1 87 pg/mL-120 pg/mL Sensitivity 0 47 pg/mL Sample serum plasma tissue homogenates Method Sandwich