Learn More
Biomatik Corporation Rat Suppressors Of Cytokine Signaling 2 (SOCS2) ELISA Kit
Supplier:  Biomatik Corporation EKU07515
Encompass_Preferred
Rat Suppressors Of Cytokine Signaling 2 (SOCS2) ELISA Kit for detecting SOCS2 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88582. Gene ID: 84607. Target Synonyms: CIS2; Cish2; SOCS-2; SSI-2; SSI2; STATI2; Cytokine-inducible SH2 protein 2; STAT-induced STAT inhibitor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6637