Learn More
Biomatik Corporation Rat Suppressors Of Cytokine Signaling 1 (SOCS1) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11253
Encompass_Preferred
Rat Suppressors Of Cytokine Signaling 1 (SOCS1) Mini sample ELISA Kit for detecting SOCS1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QX78. Target Synonyms: CIS1; CISH1; JAB; SOCS-1; SSI-1; SSI1; TIP3; JAK-binding protein; STAT-induced STAT inhibitor 1; Tec-interacting protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8183