Learn More
Biomatik Corporation Rat Suprabasin (SBSN) ELISA Kit
Supplier:  Biomatik Corporation EKU12064
Encompass_Preferred
Rat Suprabasin (SBSN) ELISA Kit for detecting SBSN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F7FEM5. Gene ID: 292793. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5186