Learn More
Biomatik Corporation Rat Synaptophysin (SYP) ELISA Kit
Supplier:  Biomatik Corporation EKU10235
Encompass_Preferred
Rat Synaptophysin (SYP) ELISA Kit for detecting SYP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07825. Gene ID: 24804. Target Synonyms: Major synaptic vesicle protein p38. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7277