Learn More
BIOHIPPO Rat SYP Microsample
SDP
Catalog No. 5000315636
Encompass
Rat SYP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SYP in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 05 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat SYP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat SYP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat SYP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15636 Supplier BIOHIPPO Supplier No. BHE15212478
Rat SYP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SYP in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 05 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat SYP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat SYP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat SYP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color