Learn More
Biomatik Corporation Rat Tachykinin Receptor 1 (TACR1) ELISA Kit
Supplier:  Biomatik Corporation EKU12452
Encompass_Preferred
Rat Tachykinin Receptor 1 (TACR1) ELISA Kit for detecting TACR1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14600. Gene ID: 24807. Target Synonyms: NK1R; SPR; NKIR; TAC1R; Neurokinin 1 Receptor; Substance P Receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6483