Learn More
BIOHIPPO Rat TF Microsample
SDP
Catalog No. 5000315318
Encompass
Rat TF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TF in plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 13 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat TF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat TF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat TF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15318 Supplier BIOHIPPO Supplier No. BHE15211981
Rat TF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TF in plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 13 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat TF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat TF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat TF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color