Learn More
Abcam Recombinant Rat TGF beta 1 protein (Tagged), 100UG
Supplier:  Abcam AB236341100UG
Encompass_Preferred
Recombinant Rat TGF beta 1 protein (Tagged) is a Rat Fragment protein, in the 30 to 278 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Transforming growth factor beta-1 proprotein, Tgfb1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1811382