Learn More
BIOHIPPO Rat TGFbR2 Microsample
SDP
Catalog No. 5000314653
Encompass
Rat TGFbR2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TGFbR2 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 126 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat TGFbR2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat TGFbR2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat TGFbR2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14653 Supplier BIOHIPPO Supplier No. BHE15210795
Rat TGFbR2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TGFbR2 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 126 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat TGFbR2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat TGFbR2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat TGFbR2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color