Learn More
BIOHIPPO Rat TH Microsample
SDP
Catalog No. 5000315786
Encompass
Rat TH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TH in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat TH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat TH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat TH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15786 Supplier BIOHIPPO Supplier No. BHE15211223
Rat TH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TH in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat TH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat TH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat TH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color