Learn More
Sino Biological Anti-Thrombomodulin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80348T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Thrombomodulin / THBD (Catalog#80348-R08H; O35370; Met1-Ser517). Thrombomodulin / THBD specific IgG was purified by Rat Thrombomodulin / THBD affinity chromatography.
Catalog No. 50-255-2777