Learn More
Immunology Consultants Rat Thiostatin ELISA Kit
SDP
Supplier:  Immunology Consultants E25TST
Encompass_Preferred
The Rat Thiostatin (TST) test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring TST in rat biological samples.
Catalog No. 50-240-6573