Learn More
Biomatik Corporation Rat Thrombospondin 4 (THBS4) ELISA Kit
Supplier:  Biomatik Corporation EKU11470
Encompass_Preferred
Rat Thrombospondin 4 (THBS4) ELISA Kit for detecting THBS4 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49744. Target Synonyms: TSP4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6268