Learn More
American Research Products Inc Rat Thyroid Stimulating Hormon
Supplier:  American Research Products Inc CSBE05115R
Encompass
Rat Thyroid Stimulating Hormone TSH ELISA Kit from CUSABIO Detection Range 0 6 IU/mL-24 IU/mL Sensitivity 0 3 IU/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26555