Learn More
Biomatik Corporation Rat Thyroid Stimulating Hormone Receptor (TSHR) ELISA Kit
Supplier:  Biomatik Corporation EKU07685
Encompass_Preferred
Rat Thyroid Stimulating Hormone Receptor (TSHR) ELISA Kit for detecting TSHR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21463. Gene ID: 25360. Target Synonyms: LGR3; HTSHR-I; Thyrotropin Receptor; TSH Receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8735