Learn More
Biomatik Corporation Rat Thyroxine Binding Globulin (TBG) ELISA Kit
Supplier:  Biomatik Corporation EKU11883
Encompass_Preferred
Rat Thyroxine Binding Globulin (TBG) ELISA Kit for detecting TBG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.37ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35577. Target Synonyms: SERPINA7; Serpin Peptidase Inhibitor Clade A Member 7; T4-binding globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8069