Learn More
Biomatik Corporation Rat Thyroxine, T4 Elisa Kit
Supplier:  Biomatik Corporation EKC31499
Encompass_Preferred
Rat Thyroxine, T4 Elisa Kit for detecting Thyroxine, T4 in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 20-320ng/ml. Sensitivity: 20ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6129