Learn More
Biomatik Corporation Rat Tight Junction Associated Protein 1, Peripheral (TJAP1) ELISA Kit
Supplier:  Biomatik Corporation EKU11471
Encompass_Preferred
Rat Tight Junction Associated Protein 1, Peripheral (TJAP1) ELISA Kit for detecting TJAP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5,000pg/ml. Sensitivity: 34pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZU31. Gene ID: 316233. Target Synonyms: PILT; TJP4; Tight Junction Protein 4; Protein incorporated later into tight junctions. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5786