Learn More
American Research Products Inc Rat tight junction protein 1
Catalog No. 5000328517
Encompass
Rat tight junction protein 1 (ZO1) ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28517 Supplier American Research Products Inc Supplier No. CSBE17287R
Rat tight junction protein 1 (ZO1) ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich