Learn More
Biomatik Corporation Rat Tight Junction Protein 3 (TJP3) ELISA Kit
Supplier:  Biomatik Corporation EKU11472
Encompass_Preferred
Rat Tight Junction Protein 3 (TJP3) ELISA Kit for detecting TJP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.47-30ng/ml. Sensitivity: 0.19ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3Z8G7. Target Synonyms: ZO-3; Zona Occludens 3; Zonula occludens protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8137