Learn More
Sino Biological Recombinant Rat TIGIT Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 8A6318R02H100
Encompass_Preferred
A DNA sequence encoding the rat TIGIT (XP_008766987.1) (Met1-Ala138) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-2752