Learn More
Raybiotech Inc Rat TIMP-1 ELISA Kit
Supplier:  Raybiotech Inc ELRTIMP1CL1
Encompass_Preferred
For cell and tissue lysate samples. Sensitivity: 4 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes rat TIMP-1.
Catalog No. 50-195-0308