Learn More
Sino Biological Anti-TIMP-1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80064MM12100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat TIMP-1 (rR TIMP-1; Catalog# NP_446271.1; P01860; Met1-Ala217). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-2842