Learn More
Sino Biological Recombinant Anti-TIMP-1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80064R009100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat TIMP-1 (rR TIMP-1; Catalog#80064-RNAH; NP_446271.1; Met1-Ala217).
Catalog No. 50-255-2841