Learn More
American Research Products Inc Rat Tissue factor pathway inhi
Catalog No. 5000329668
Encompass
Rat Tissue factor pathway inhibitor TFPI ELISA Kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29668 Supplier American Research Products Inc Supplier No. CSBE08261R
Rat Tissue factor pathway inhibitor TFPI ELISA Kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich