Learn More
American Research Products Inc Rat tissue factor TF coagul
Catalog No. 5000327372
Encompass
Rat tissue factor (TF) /coagulation factor VII (F7) compleX ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27372 Supplier American Research Products Inc Supplier No. CSBEQ027252RA
Rat tissue factor (TF) /coagulation factor VII (F7) compleX ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich