Learn More
American Research Products Inc Rat tissue inhibitors of metal
Catalog No. 5000328892
Encompass
Rat tissue inhibitors of metalloproteinase 1 TIMP-1 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 224 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28892 Supplier American Research Products Inc Supplier No. CSBE08005R
Rat tissue inhibitors of metalloproteinase 1 TIMP-1 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 224 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich