Learn More
American Research Products Inc Rat tissue inhibitors of metal
Catalog No. 5000326442
Encompass
Rat tissue inhibitors of metalloproteinase 2 TIMP-2 ELISA Kit from CUSABIO Detection Range 1 25 ng/mL-80 ng/mL Sensitivity 0 31 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26442 Supplier American Research Products Inc Supplier No. CSBE07386R
Rat tissue inhibitors of metalloproteinase 2 TIMP-2 ELISA Kit from CUSABIO Detection Range 1 25 ng/mL-80 ng/mL Sensitivity 0 31 ng/mL Sample serum plasma tissue homogenates Method Sandwich