Learn More
ACROBiosystems Rat TL1A Protein His Tag
Supplier:  ACROBiosystems TLAR5249100UG
Encompass_Preferred
Rat TL1A Protein His Tag (TLA-R5249) is expressed from human 293 cells (HEK293). It contains AA Thr 61 - Ile 252 (Accession # Q8K3Y7).
Catalog No. 50-253-4036