Learn More
Sino Biological Anti-TNF-alpha Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80045T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat TNF-alpha/TNFA/TNFSF2 (rR TNF-alpha/TNFA/TNFSF2; Catalog#80045-RNAE; P16599; Leu80-Leu235). TNF-alpha/TNFA/TNFSF2 specific IgG was purified by Rat TNF-alpha/TNFA/TNFSF2 affinity chromatography.
Catalog No. 50-240-8956