Learn More
Raybiotech Inc Rat TNF RI (TNFRSF1A) ELISA Kit
Supplier:  Raybiotech Inc ELRTNFR11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 2 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects rat TNF R1. Other species not determined.
Catalog No. 50-195-0312