Learn More
Sino Biological Anti-RANK/TNFRSF11A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80160RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat TNFRSF11A (rR TNFRSF11A; Catalog#80160-R08H; XP_573424.2; Met1-Pro213). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2818