Learn More
Sino Biological Anti-RANK/TNFRSF11A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80160T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat TNFRSF11A (Catalog#80160-R08H; XP_573424.2; Met1-Pro213). TNFRSF11A specific IgG was purified by Rat TNFRSF11A affinity chromatography.
Catalog No. 50-255-2817