Learn More
Sino Biological Recombinant Rat TL1A/TNFSF15 Protein (Trimer, His Tag), MALS-verified
Catalog No. 502992732
Encompass_Preferred
A DNA sequence encoding the Rat TNFSF15 (Q8K3Y7) (Val76-Ile252) was expressed, with a polyhistidine tag at the N-terminus. This construct has a trimer design.
Catalog No. 50-299-2732 Supplier Sino Biological Supplier No. 80150R07H1
A DNA sequence encoding the Rat TNFSF15 (Q8K3Y7) (Val76-Ile252) was expressed, with a polyhistidine tag at the N-terminus. This construct has a trimer design.